Eisen ≥99,995% (Metall-Basis), Folie, Puratronic®, Dicke 0.25 mm (0.01 in)
Lieferant: Thermo Fisher Scientific
11380.FF
11380.FI
11380.GH
11380.FFEA
81.8
EUR
11380.FF
11380.FI
11380.GH
Eisen ≥99,995% (Metall-Basis), Folie, Puratronic®, Dicke 0.25 mm (0.01 in)
Iron
Purity: 99.995%
CAS No.: 7439-89-6
Physical Form: ≈1.23g/25x25mm
MDL No.: MFCD00010999
Formel:
Fe Siedepunkt: 3000 °C (1013 hPa) Schmelzpunkt: 1535 °C Dichte: 7,86 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010999 CAS-Nummer: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...