Di-sec-butoxyaluminoxytriethoxysilane

Lieferant: Thermo Fisher Scientific
Achtung

41210.09 41210.18 41210.30
41210.09EA 49.8 EUR
41210.09 41210.18 41210.30
Di-sec-butoxyaluminoxytriethoxysilane
Di-sec-butoxyaluminoxytriethoxysilane

CAS No.: 68959-06-8 Molecular Formula: C14H33AlO6Si Formula Weight: 352.48 Flash Point: 46°(115°F) Physical Form: Liquid Density: 1.00 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 200° UN No.: UN1993 MDL No.: MFCD00053771

Formel: C₁₄H₃₃AlO₆Si
Siedepunkt: >200 °C
Dichte: 1,00
Flammpunkt: 46 °C (115 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00053771
CAS-Nummer: 68959-06-8
EINECS: 273-392-6
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR