Copper(II) methacrylate, tech.

Lieferant: Thermo Fisher Scientific
Achtung

41621.06 41621.14
41621.06EA 58 EUR
41621.06 41621.14
Copper(II) methacrylate, tech.
Copper(II) methacrylate

CAS No.: 19662-59-0 Molecular Formula: C8H10CuO4 Formula Weight: 233.71 Physical Form: Powder Appearance: Dark blue-green MDL No.: MFCD00080547

Formel: C₈H₁₀CuO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00080547
CAS-Nummer: 19662-59-0
EINECS: 243-211-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR