4-Chlor-DL-phenylalanin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH

A13323.09 A13323.18
A13323.09EA 89 EUR
A13323.09 A13323.18
4-Chlor-DL-phenylalanin ≥98%
4-Chlor-DL-phenylalanin

Purity: 98+% CAS No.: 7424-00-2 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Melting Point: ca 265° dec. Beilstein Reference: 2805758

Formel: C₉H₁₀ClNO₂
Schmelzpunkt: ∼ 265 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002601
CAS-Nummer: 7424-00-2
EINECS: 231-051-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR