Anti-MFGE8 Mouse Monoclonal Antibody [clone: MFG-06]

Lieferant: Abnova
MAB3634
ABNOMAB3634EA 402 EUR
ABNOMAB3634
Anti-MFGE8 Mouse Monoclonal Antibody [clone: MFG-06]
Antikörper
Mouse monoclonal antibody raised against native MFGE8.

Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: MFG-06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR