Anti-Dinitrophenyl Hapten Monoclonal Antibody [clone: LO-DNP-2]

Lieferant: Abnova
MAB5195
ABNOMAB5195EA 1170 EUR
ABNOMAB5195
Anti-Dinitrophenyl Hapten Monoclonal Antibody [clone: LO-DNP-2]
Antikörper
Rat monoclonal antibody raised against Dinitrophenyl Hapten.

Type: Primary
Antigen: Dinitrophenyl Hapten
Clonality: Monoclonal
Clone: LO-DNP-2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR