Zinn ACS 20 mesh
Lieferant: Thermo Fisher Scientific
420985000
420981000
ACRO420985000EA
181
EUR
ACRO420985000
ACRO420981000
Zinn ACS 20 mesh
Zinn
Formel:
Sn Siedepunkt: 2270 °C (760 mmHg) Schmelzpunkt: 231,9 °C Dichte: 7,31 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00133862 CAS-Nummer: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Spezifikation Testergebnisse
Appearance | Grey Metallic granular powder |
Titration Complexometric | ≥99.5 % |
Antimony (Sb) | ≤200 ppm |
Arsenic (As) | ≤1 ppm |
Copper (Cu) | ≤50 ppm |
Iron (Fe) | ≤100 ppm |
Lead (Pb) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...