N-(2-Propynyl)-2,3-dihydroinden-1-amine 95%

Lieferant: Apollo Scientific

OR965634-1G
APOSOR965634-1GEA 111 EUR
APOSOR965634-1G
N-(2-Propynyl)-2,3-dihydroinden-1-amine 95%
N-(2-Propynyl)-2,3-dihydroinden-1-amine
Formel: C₁₂H₁₃N
Molecular Weight: 171,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00033000
CAS-Nummer: 1875-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR