2-(Trifluormethoxy)benzylamin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2-(Trifluoromethoxy)phenyl)methanamine
PC7438NP-1G
PC7438NP-25G
PC7438NP-5G
APOSPC7438NP-1GEA
21.9
EUR
APOSPC7438NP-1G
APOSPC7438NP-25G
APOSPC7438NP-5G
2-(Trifluormethoxy)benzylamin 95%
2-(Trifluormethoxy)benzylamin
Formel:
C₈H₈F₃NO Siedepunkt: 77 °C/12mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085176 CAS-Nummer: 175205-64-8 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...