3,4-Difluorophenylacetyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC9314-1G
PC9314-250MG
APOSPC9314-1GEA
188
EUR
APOSPC9314-1G
APOSPC9314-250MG
3,4-Difluorophenylacetyl chloride
3,4-Difluorophenylacetyl chloride
Formel:
C₈H₅ClF₂O Siedepunkt: 91…92 °C (6 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458138 CAS-Nummer: 121639-61-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...