Anti-STOML2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-8861R-A680
BOSSBS-8861R-A680EA 893 EUR
BOSSBS-8861R-A680
Anti-STOML2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
STOML2 is a 356 amino acid member of the mec-2 family of proteins. Expressed ubiquitously at low levels, STOML2 is highly expressed in heart, liver and pancreas. STOML2 is localised to the cytoplasm with some distribution on the membrane. STOML2 was first identified as an overexpressed protein in human endometrial adenocarcinoma. Changes in cell growth in samples with different levels of STOML2 indicate that STOML2 could play a role in endometrial tumorigenesis. STOML2 is also thought to play a role in regulating ion channel conductances or the organisation of sphingolipid and cholesterol-rich lipid rafts.

Type: Primary
Antigen: STOML2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-356/356
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR