1-Boc-4-(hydroxymethyl)piperidin ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

H26650.03 H26650.06
H26650.03EA 34.3 EUR
H26650.03 H26650.06
1-Boc-4-(hydroxymethyl)piperidin ≥97%
1-Boc-4-(hydroxymethyl)piperidin

Purity: 97% CAS No.: 123855-51-6 Molecular Formula: C11H21NO3 Formula Weight: 215.29 Melting Point: 78-82° MDL No.: MFCD02094488

Formel: C₁₁H₂₁NO₃
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02094488
CAS-Nummer: 123855-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR