1-Hexansulfonsäure Natriumsalz, weiße Flocken

Lieferant: MP Biomedicals

Synonyme: Natriumhexylsulfonat

0215125201 0215125205 0215125210
ICNA0215125201EA 22.7 EUR
ICNA0215125201 ICNA0215125205 ICNA0215125210
1-Hexansulfonsäure Natriumsalz, weiße Flocken
1-Hexansulfonsäure Natriumsalz

Sodium hexanesulfonate is a low molecular weight alkylsulfonate that is used as an ion pairing reagent for HPLC and as an anionic surfactant. The anionic sulfonate counterion permits the separation and resolution of positively charged analytes. Because of its low molecular weight, it will not form micelles in solution and therefore has no CMC value.
For ion-pair chromatography. Sodium hexanesulfonate is utilized in the analysis of small organic compounds, and pharmaceutical products and metabolites by such techniques as HPLC.


  • Presentation: White Flakes
  • Soluble in water (100 mg/ml-clear, colorless), ethanol or methanol
  • Storage temperature: Room Temperature

Formel: C₆H₁₃O₃SNa
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007542
CAS-Nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Reference
Moisture ≤10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR