7-Indolcarbonsäure

Lieferant: Molekula

Synonyme: 1H-Indole-7-carboxylic acid

M53645945
MOLEM53645945EA 0 EUR
MOLEM53645945
7-Indolcarbonsäure
7-Indolcarbonsäure
Formel: C₉H₇NO₂
MDL: MFCD00210442
CAS-Nummer: 1670-83-3
EINECS: 216-801-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR