Anti-SHPK Mouse Monoclonal Antibody [clone: OTI3D7]

Lieferant: OriGene
TA503210
ORIGTA503210EA 621 EUR
ORIGTA503210
Anti-SHPK Mouse Monoclonal Antibody [clone: OTI3D7]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: SHPK (sedoheptulokinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR