3-(Trimethylsilyl)-2-propin-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Trimethylsilyl)propargyl alcohol

M-Clarity™ Qualitätsstufe = MQ100

318558-5G
SIAL318558-5GEA 59.6 EUR
SIAL318558-5G
3-(Trimethylsilyl)-2-propin-1-ol, Sigma-Aldrich®
3-(Trimethylsilyl)-2-propin-1-ol
Formel: C₆H₁₂OSi
Molecular Weight: 128,25 g/mol
MDL: MFCD00002913
CAS-Nummer: 5272-36-6
EINECS: 226-094-5
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR