3',5'-Dinitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3,5-Dinitrophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

631299-1G
SIAL631299-1GEA 113 EUR
SIAL631299-1G
3',5'-Dinitroacetophenon, Sigma-Aldrich®
3',5'-Dinitroacetophenon
Formel: C₈H₆N₂O₅
MDL: MFCD00631987
CAS-Nummer: 14401-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR