1,1-Dimethylbiguanid Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

D150959-5G
SIALD150959-5GEA 43.4 EUR
SIALD150959-5G
1,1-Dimethylbiguanid Hydrochlorid, Sigma-Aldrich®
1,1-Dimethylbiguanid Hydrochlorid
Formel: C₄H₁₂ClN₅
MDL: MFCD00012582
CAS-Nummer: 1115-70-4
EINECS: 214-230-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR