Anti-Amino Acid Arylamidase Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
A1372-95B
USBIA1372-95BEA 626 EUR
USBIA1372-95B
Anti-Amino Acid Arylamidase Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Amino Acid Arylamidase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR