Anti-Milk Fat Globulin Mouse Monoclonal Antibody [clone: MFG-06]
Lieferant: US Biological
M3897-08B
USBIM3897-08BEA 608 EUR
USBIM3897-08B
Anti-Milk Fat Globulin Mouse Monoclonal Antibody [clone: MFG-06]
Antikörper
Type: Primary
Antigen: Milk Fat Globulin
Clonality: monoclonal
Clone: MFG-06
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human