Anti-MBOAT7 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
AVIVARP49810_P050EA 385 EUR
AVIVARP49810_P050
Anti-MBOAT7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MBOAT7 (membrane bound O-acyltransferase domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Rat