Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)

Lieferant: Biotium

20016-1EA 157 EUR
20016-1 20016.
Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR