Anti-IgG Antibody (Cy3®)

Lieferant: Boster
BSBTBA1034EA 239 EUR
BSBTBA1034
Anti-IgG Antibody (Cy3®)
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR