Anti- IgG Antibody (Unconjugated)

Lieferant: Boster
BSBTBA1044-5EA 0 EUR
BSBTBA1044-5
Anti- IgG Antibody (Unconjugated)
Antikörper
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR