Anti-IgA Antibody (HRP (Horseradish Peroxidase))

Lieferant: Boster
BSBTBA1066-0.25EA 0 EUR
BSBTBA1066-0.25
Anti-IgA Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgA
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR