Anti-Avidin Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Boster
BSBTBA1125-0.5EA 0 EUR
BSBTBA1125-0.5
Anti-Avidin Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Primary
Antigen: Avidin
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR