N-Methoxy-N-methylbenzamide 98%

Lieferant: Thermo Scientific
Achtung

ACRO436560050EA 77.3 EUR
ACRO436560050
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Siedepunkt: 85 °C (1 mmHg)
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075320
CAS-Nummer: 6919-61-5

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR