Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: TR3.06]

Lieferant: ProSci
PRSI36-139EA 612 EUR
PRSI36-139
Anti-TNFRSF10C Mouse Monoclonal Antibody [clone: TR3.06]
Antikörper
TRAIL-R3 is a receptor for the cytotoxic ligand TRAIL. It lacks a cytoplasmic death domain and hence is not capable of inducing apoptosis. Protects cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.

Immunohistochemistry: (paraffin sections) (20ug/ml). Western Blot: (1ug/ml) . Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF10C
Clonality: Monoclonal
Clone: TR3.06
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR