Anti-TNFRSF13C Rat Monoclonal Antibody [clone: 9B9]
Lieferant: ProSci
PRSI36-141EA 584 EUR
PRSI36-141
Anti-TNFRSF13C Rat Monoclonal Antibody [clone: 9B9]
Antikörper
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Functional Application: Depletion of B cells in vivo. Optimal conditions must be determined individually for each application.
Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: 9B9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse