Anti-TNFSF13B Rat Monoclonal Antibody [clone: 1-35-1]

Lieferant: ProSci
PRSI36-149EA 565 EUR
PRSI36-149
Anti-TNFSF13B Rat Monoclonal Antibody [clone: 1-35-1]
Antikörper
BAFF is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation. BAFF co-stimulates activated T cells. Increased levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren’s syndrome, rheumatoid arthritis, multiple sclerosis and systemic lupus erythematosus (SLE). Furthermore, BAFF is found in inflammatory sites in which there is lymphoid neogenesis. BAFF levels are elevated in patients with multiple myeloma and B cell chronic lymphoid leukemia (B-CCL).

Type: Primary
Antigen: TNFSF13B
Clonality: Monoclonal
Clone: 1-35-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR