Anti-Bid Polyclonal Antibody

Lieferant: Boster
BSBTPB9027EA 550 EUR
BSBTPB9027
Anti-Bid Polyclonal Antibody
Antikörper
Polyclonal antibody for BID detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. BID information: Molecular Weight: 30644 MW; Subcellular Localization: Secreted. The lack of a specific hydrophobic segment in the precursor sequence suggests that IL-1 is released by damaged cells or is secreted by a mechanism differing from that used for other secretory proteins; Tissue Specificity: Isoform 2 and isoform 3 are expressed in spleen, bone marrow, cerebral and cerebellar cortex. Isoform 2 is expressed in spleen, pancreas and placenta (at protein level). Isoform 3 is expressed in lung, pancreas and spleen (at protein level). Isoform 4 is expressed in lung and pancreas (at protein level).

Type: Primary
Antigen: Bid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR