Cleanroom wipes, C3
Lieferant: FG Cleanwipes
EPTA7-C3-99L-00EA 1130 EUR
EPTA7-C3-99L-00
Cleanroom wipes, C3
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
These 100% hydroentangled polyester wipes offer the highest level of softness and abrasion resistance − perfect for cleaning and polishing the most sensitive surfaces.
  • Tough and absorbent for usage in a variety of applications
  • Recommended for the cleaning of lenses or other sensitive surfaces
  • Chemically compatible with most cleaning solutions
  • Low NVRs and ions
  • No chemical binders
Typical applications include: Optics cleaning and manufacturing, LCD and display cleaning and maintenance, sensitive surface wipe downs and prep, use as a work surface for delicate tasks, critical tool and component wipe downs.
Recommended for use in ISO 5 - 8 controlled environments.