7-Methyl-1H-indole-3-carboxylic acid 98%
Lieferant: Apollo Scientific
OR913136-5G
APOSOR913136-5GEA
55.1
EUR
APOSOR913136-5G
7-Methyl-1H-indole-3-carboxylic acid 98%
7-Methyl-1H-indole-3-carboxylic acid
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,1800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203634 CAS-Nummer: 30448-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...