X-MINI® pressor, Corning®
Lieferant: Corning
Corning®
734-3398EA 3230 EUR
734-3398
X-MINI® pressor, Corning®
Bioreaktoren
The Corning X-MINI Pressor is designed to be used exclusively with the Corning X-MINI Selection Kits.
The Corning X-MINI Pressor allows for the degassing of microbubble-targeted cells. It accommodates a capped 10 ml syringe.
The pressure generated using the X-MINI Pressor is sufficient to isolate up to 1×10⁷ cells, and one syringe can process up to 3 ml of cell suspension at a time.