Anti-TA-650 ICM3 Recombinant Antibody

Lieferant: ProSci
PRSI10-176EA 627 EUR
PRSI10-176
Anti-TA-650 ICM3 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR