Methylcellulose, 1500 mPa.S (2% H₂O) USP, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ400
SIALM0555-500GEA
759
EUR
SIALM0555-500G
SIALM0555-100G
Methylcellulose, 1500 mPa.S (2% H₂O) USP, Sigma-Aldrich®
Methyl cellulose
1 mPa.s = 1cP
Standards: Meets USP testing specifications.
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00081763 CAS-Nummer: 9004-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...