2,6-Dichlornicotinonitril, Sigma-Aldrich®

Lieferant: Merck

SIAL703079-1GEA 34.6 EUR
SIAL703079-1G
2,6-Dichlornicotinonitril, Sigma-Aldrich®
2,6-Dichlornicotinonitril
Formel: C₆H₂Cl₂N₂
Molecular Weight: 173,00 g/mol
MDL: MFCD03411341
CAS-Nummer: 40381-90-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR