Dimethylsulfoxid ≥99,99995% (Metall-Basis) für anorganische Spurenanalyse, Supelco®
Lieferant: Merck
Synonyme:
Methylsulfinylmethan, DMSO
01934-1L
01934-1LEA
214
EUR
01934-1L
Dimethylsulfoxid ≥99,99995% (Metall-Basis) für anorganische Spurenanalyse, Supelco®
Dimethylsulfoxid
Bitte beachten Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.
Formel:
(CH₃)₂SO Molecular Weight: 78,14 g/mol |
MDL:
MFCD00002089 CAS-Nummer: 67-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...