Natriumnitrit, BAKER, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Diazotizing salts

0298.1000EA 72.55 EUR
0298.1000
Natriumnitrit, BAKER, Laborreagens, J.T.Baker®
Sodium nitrite
Formel: NaNO₂
Molecular Weight: 69 g/mol
Siedepunkt: 320 °C (1013 hPa)
Schmelzpunkt: 284 °C
Dichte: 2,168 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011118
CAS-Nummer: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721

Bestellen

Spezifikation Testergebnisse

Assay (dried basis) 98.5 - 100.5 %
Acidity or Alkalinity passes test
Appearance of solution passes test
Chloride (Cl) max. 50 ppm
Heavy Metals (as Pb) max. 20 ppm
Identification passes test
Loss on Drying max. 0.25 %
Sulfate (SO4) max. 200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR