Hämatoxylin in wässriger Lösung according to Delafield für die Mikroskopie

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DcH-48, Natural Black I, Hydroxybrasilin

M-Clarity™ Qualitätsstufe = MQ100

03971-250ML
03971-250MLEA 67.4 EUR
03971-250ML
Hämatoxylin in wässriger Lösung according to Delafield für die Mikroskopie
Haematoxylin staining solutions

Hematoxylin solution according to Delafield is a naturally ripened alum hematoxylin with potassium alum or ammonium alum as the mordant. It is used when the counterstain doesn′t possess any acid content in it. It is suitable to be used as a nuclear stain with eosin.


Hematoxylin solution according to Delafield has been used to stain the nuclei of different sections of adipose tissue for its histochemical analysis. It is suitable for staining thick or thin blood films. It is also frequently used to stain and study the visibility of microfilariae.

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Dichte: 0,950…1,044 g/ml (20 °C)
Flammpunkt: 42 °C (geschlossener Tiegel)
Lagertemperatur: Room Temperature
MDL: MFCD00078111
CAS-Nummer: 517-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR