Penicillin : Streptomycin-Lösung (20,000 IU Pen/mL, 20,000 µg Strep/mL), steril

Lieferant: MP Biomedicals
Gefahr

091670454
091670454.EA 177 EUR
091670454.
Penicillin : Streptomycin-Lösung (20,000 IU Pen/mL, 20,000 µg Strep/mL), steril
Penicillin : Streptomycin

Double antibiotic solution containing 20,000 units/mL of penicillin and 20,000 µg/mL of streptomycin.


  • Dual antibiotic solution
  • Supplements cell culture media
  • Effective against most gram positive and gram negative bacteria


Penicillin-Streptomycin is is a dual antibiotic solution used to supplement cell culture media to control bacterial contamination. The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. This solution contains 17,000 - 21,000 units/mL of penicillin and 18.0 - 23.0 mg/mL of streptomycin.


Bitte beachten For laboratory use. Not for injection.

Lagertemperatur: Gefrierschrank
CAS-Nummer: 113-98-4 (Penicillin G potassium salt) - 5490-27-7 (Dihydrostreptomycin sulfate)

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR