Schiff-Reagenz stark, Sigma-Aldrich®
Lieferant: Merck
Achtung
1.02572.1000
1.02572.1000EA 91.7 EUR
1.02572.1000
Schiff-Reagenz stark, Sigma-Aldrich®
Schiff-Reagenz
For the detection of aldehydes, polysaccharides (glycogen), and mucosubstances (glycoproteins, glycolipids, mucins)
The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for the detection of aldehyde and mucosubstances in microscopy. Schiff’s reagent intense is a ready to use, high capacity staining solution (up to 3000 slides/500 ml). It can be stored at room temperature which allows for 10 minutes of time saving per staining procedure. This new formulation provides for more intense staining results with clearer differentiation between cell types.