Kollodium (Nitrocellulose in einer Mischung aus Ethanol und Diethylether) 4%, SAFC®

Lieferant: Merck
Gefahr

1.02644.1000
1.02644.1000EA 266 EUR
1.02644.1000
Kollodium (Nitrocellulose in einer Mischung aus Ethanol und Diethylether) 4%, SAFC®
Kollodium (Nitrocellulose in einer Mischung aus Ethanol und Diethylether)

Collodium 4% (solution of cellulose nitrate) has a nitrogen content of not more than 12,6% with respect to the dry matter of cellulose nitrate.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Dichte: 0,76 g/cm³ (20 °C)
Flammpunkt: −40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081525
CAS-Nummer: 9004-70-0
UN: 2059
ADR: 3,I

Bestellen

Spezifikation Testergebnisse

Explosion Limit 1,7 - 36% (V)
Ignition Temperature 170 °C
Vapor Pressure 576 hPa (20 °C)
Storage Store at 15 to 25 °C
Appearance Colorless to Faint Yellowish Clear to Opalescent Viscous Liquid
Assay 4,0 - 4,2%
Identity (IR-spectrum) Passes Test
Neutral Reaction Passes Test
Density (d 20 °C/ 4 °C) 0,750 - 0,770 g/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR