3,3'-Diaminobenzidin, Tabletten Puffersubstanz, für die Färbung in der Mikroskopie, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: DAB, Biphenyl-3,3',4,4'-tetrayltetraamin

1.02924.0001
1.02924.0001EA 207 EUR
1.02924.0001
3,3'-Diaminobenzidin, Tabletten Puffersubstanz, für die Färbung in der Mikroskopie, Sigma-Aldrich®
3,3'-Diaminobenzidin

Peroxidase chromogen for immunohistochemistry (IHC)


3,3'-Diaminobenzidine (DAB) is a peroxidase substrate producing a brown colored product that is insoluble in alcohol. The Peroxidase chromogen is used for the visualisation of antigenic structures in cryostat and paraffin sections in immunohistochemistry, and should be used at non-aqueous specimen.The reagent is sufficient for 50 staining procedures. It is registered as IVD and CE product.

Formel: C₁₂H₁₄N₄
Molecular Weight: 214,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00007725
CAS-Nummer: 91-95-2
IVDLogo

Bestellen

Spezifikation Testergebnisse

Suitability for microscopy passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR