Ion exchange chromatography media, Ion Exchanger I
Lieferant: Merck
1.04765.0500
1.04765.0500EA
155
EUR
1.04765.0500
Ion exchange chromatography media, Ion Exchanger I
Protein-Aufbereitungs
Protein Purification Resins
Ion Exchanger I is a strong acid cation exchangers widely used in separation of noble earths and amino acids, water treatment, analysis and food industry. The bond strength of equivalent ions increases with decreasing diameter of the hydrated ion.
Maximum working temperature: 120 °C
Exchange capacity: >1,7 mval/ml