Dichlormethan ≥99,9% (durch GC) stabilisiert, Suprasolv® hypergrade für die organische Spurenanalyse, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
Methylenchlorid, Chlormethylen
Stabilisierung:
Stabilisiert mit 2-Methyl-2-buten (iso-Amylen)
M-Clarity™ Qualitätsstufe = MQ100
1.06454.1000
1.06454.1000EA
189
EUR
1.06454.1000
Dichlormethan ≥99,9% (durch GC) stabilisiert, Suprasolv® hypergrade für die organische Spurenanalyse, Supelco®
Dichlormethan
Suprasolv® hypergrade is a unique and universal solvent quality which can be employed for all three GC detection methods (ECD, FID and MS). Thanks to the large retention time range, even low-boiling substances can be reliably analyzed.
Formel:
CH₂Cl₂ Molecular Weight: 84,93 g/mol Siedepunkt: 39,8 °C (1013 hPa) Schmelzpunkt: –95 °C Dichte: 1,322 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000881 CAS-Nummer: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 12,06140 |
Spezifikation Testergebnisse
Purity (GC) | ≥ 99,9% |
Identity (IR) | conforms |
Evaporation Residue | ≤3,0 mg/l |
Water | ≤0,005% |
Colour | ≤10 Hazen |
GC/ECD (Retention Range 1,2,4-Tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)) | ≤2 pg/ml |
GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)) | ≤2 ng/ml |
GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning Area 30 - 600 amu, Individual Signals (n-Tetradecane Standard)) | ≤2 ng/ml |
Recommended for Analysis of Polychlorinated Dibenzodioxins and Polychlorinated Dibenzofurans (PCDD/F) | |
The Product is Stabilised with 2-Methyl-2-Butene |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...