Xylol (Isomerengemisch) für die Histologie
Lieferant: Merck
|
Gefahr
|
Synonyme:
Dimethylbenzol
1.08298.4000
1.08298.4000EA
160
EUR
1.08298.4000
Xylol (Isomerengemisch) für die Histologie
Xylol (Isomerengemisch)
Histological grade Xylene was developed to be a robust clearing agent in both histological and cytological applications. It is a critical raw material in all histological staining procedures. It supports the QA process in Pathology. It is a CE-certified, IVD product.
Formel:
C₆H₄(CH₃)₂ Molecular Weight: 106,17 g/mol Siedepunkt: 136…145 °C (1013 hPa) Schmelzpunkt: –25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077264 CAS-Nummer: 1330-20-7 UN: 1307 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...