Kaliumiodid-Stärke-Testpapier Reag.Ph.Eur., Supelco®
Lieferant: Merck
1.09512.0003
1.09512.0003EA 20.27 EUR
1.09512.0003
Kaliumiodid-Stärke-Testpapier Reag.Ph.Eur., Supelco®
Testpapiere Indikatorpapiere
Potassium iodine paper is used for the determination of oxidising agents.