Reflektometer, Reflectoquant® RQflex® 20
Lieferant: Merck
1.17246.0001
1.17246.0001EA 1020 EUR
1.17246.0001
Reflektometer, Reflectoquant® RQflex® 20
Reflektometer
The Reflectoquant system consists of test strips and reflectometers.
  • Intuitive menu: Dip the strip and read
  • Barcode tests with batch-specific calibration
  • Larger screen and upgraded software to guide you through the analyses
RQflex 20 gives you all the tools you need for high quality analyses for sample materials from food, water, and industrial to environmental samples. Applications are available for different matrix's from ascorbic acid in juices to HMF in honey.
Area meassured: 4×6 mm