Reagents for non Merck photometers, Spectroquant®, Supelco®
Lieferant: Merck
1.19254.0001
1.19254.0001EA 46.8 EUR
1.19254.0001
Reagents for non Merck photometers, Spectroquant®, Supelco®
Umwelt-Prüfkits
Spectroquant® test kits are suitable for use only in conjunction with Hach instruments. They do not require any further device-specific calibration.